Learn More
Medchemexpress LLC HY-111475 10mg Medchemexpress, Mitochondrial fusion promoter M1 CAS:219315-22-7 Purity:>98%
SDP
Catalog No. 502080792
Encompass_Preferred
Medchemexpress, HY-111475 10mg Mitochondrial fusion promoter M1 CAS:219315-22-7 Mitochondrial fusion promoter M1 is a mitochondrial dynamic modulator. Mitochondrial fusion promoter M1 preserves the mitochondrial function and promotes cellular respiration. Mitochondrial fusion promoter M1 alleviates cardiac and brain damage in rats with cardiac ischemia/reperfusion injury. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0792 Supplier Medchemexpress LLC Supplier No. HY11147510MG
Medchemexpress, HY-111475 10mg Mitochondrial fusion promoter M1 CAS:219315-22-7 Mitochondrial fusion promoter M1 is a mitochondrial dynamic modulator. Mitochondrial fusion promoter M1 preserves the mitochondrial function and promotes cellular respiration. Mitochondrial fusion promoter M1 alleviates cardiac and brain damage in rats with cardiac ischemia/reperfusion injury. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.