Learn More
Cayman Chemical MItochondrIal FUSN PROMTR 5mg
SDP
Catalog No. 502944103
Encompass_Preferred
A promoter of mitochondrial fusion; induces elongation of mitochondria in Mfn1-/- or Mfn2-/- MEFs (EC50s = 5.3 and 4.42 µM, respectively); protects against MPP+-induced mitochondrial fragmentation and cytotoxicity in SH-SY5Y cells at 5 µM; reduces apoptosis and inhibits decreases in testosterone levels induced by triphenyl phosphate in TM3 cells at 1 µM; improves novel object recognition deficits in a rat model of doxorubicin-induced chemobrain at 2 mg/kg
Catalog No. 50-294-4103 Supplier Cayman Chemical Supplier No. 399095 MG
A promoter of mitochondrial fusion; induces elongation of mitochondria in Mfn1-/- or Mfn2-/- MEFs (EC50s = 5.3 and 4.42 µM, respectively); protects against MPP+-induced mitochondrial fragmentation and cytotoxicity in SH-SY5Y cells at 5 µM; reduces apoptosis and inhibits decreases in testosterone levels induced by triphenyl phosphate in TM3 cells at 1 µM; improves novel object recognition deficits in a rat model of doxorubicin-induced chemobrain at 2 mg/kg