Learn More
Sigma Aldrich Fine Chemicals Biosciences Mitochondrial Membrane Pot
Catalog No. 502904939
Encompass
Mitochondria generate a potential across their membranes due to the activities of enzymes of the electron transport chain. During apoptosis collapse of the mitochondrial membrane potential (MMP) coincides with the opening of the mitochondrial permeability transition pores leading to the release of cytochrome c into the cytosol which in turn triggers other downstream events in the apoptotic cascade.
Catalog No. 50-290-4939 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. MAK1591KT
Add to Cart
Add to Cart
Mitochondria generate a potential across their membranes due to the activities of enzymes of the electron transport chain. During apoptosis collapse of the mitochondrial membrane potential (MMP) coincides with the opening of the mitochondrial permeability transition pores leading to the release of cytochrome c into the cytosol which in turn triggers other downstream events in the apoptotic cascade.