Learn More
Cayman Chemical MItochondrIal PT Pore asy K
SDP
Catalog No. 502892327
Encompass_Preferred
Determine mitochondrial inner membrane permeability through the measurement of mitochondrial membrane potential and membrane integrity
Catalog No. 50-289-2327 Supplier Cayman Chemical Supplier No. 601430100 TEST
Determine mitochondrial inner membrane permeability through the measurement of mitochondrial membrane potential and membrane integrity