Learn More
Medchemexpress LLC HY-131453A 5mg Medchemexpress, Mitochondrial respiration-IN-1 (hydrobromide) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY131453A5MG
Encompass_Preferred
Medchemexpress, HY-131453A 5mg Mitochondrial respiration-IN-1 (hydrobromide) CAS: Mitochondrial respiration-IN-1 hydrobromide (compound 49) is a potent mitochondrial inhibitor (IC50=8.8 mg/mL) extracted from patent US20110301180A1, compound 49. Mitochondrial respiration-IN-1 hydrobromide significantly reduces mitochondrial respiration in platelets. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1286