Learn More
Medchemexpress LLC Mitochondrial respiration-IN-1 hydrobromide | 00-00-0 | 99.9% | C26H26Br2NO2PS | 10MG
SDP
Catalog No. 5000259203
Encompass_Preferred
Mitochondrial respiration-IN-1 hydrobromide is a small-molecule inhibitor of mitochondrial respiration provided as the hydrobromide salt for biochemical and cellular research. It significantly reduces mitochondrial respiration in platelets and is supplied as a white to off-white solid suitable for in vitro studies.
  • High purity (99.92%).
  • Soluble in water (100 mg/mL) and DMSO (66.67 mg/mL); ultrasonic assistance may be required.
  • Stable when stored sealed away from moisture; in solution: -80°C (6 months) and -20°C (1 month).
  • White to off-white solid appearance.
  • Suitable for studies of mitochondrial function and platelet respiration.
Catalog No. 50-002-59203 Supplier Medchemexpress LLC Supplier No. HY131453A10MG
Mitochondrial respiration-IN-1 hydrobromide is a small-molecule inhibitor of mitochondrial respiration provided as the hydrobromide salt for biochemical and cellular research. It significantly reduces mitochondrial respiration in platelets and is supplied as a white to off-white solid suitable for in vitro studies.
  • High purity (99.92%).
  • Soluble in water (100 mg/mL) and DMSO (66.67 mg/mL); ultrasonic assistance may be required.
  • Stable when stored sealed away from moisture; in solution: -80°C (6 months) and -20°C (1 month).
  • White to off-white solid appearance.
  • Suitable for studies of mitochondrial function and platelet respiration.