Learn More
Medchemexpress LLC Mitochonic acid 5 | 1354707-41-7 | 99.60% | 329.30 | 100 MG
SDP
Catalog No. 5000347481
Encompass_Preferred
Mitochonic acid 5 (MA-5) is a compound that binds to mitochondria, ameliorating renal tubular and cardiac myocyte damage by modulating mitochondrial ATP synthesis independently of oxidative phosphorylation and the electron transport chain. Derived from indole-3-acetic acid, it protects mitochondrial function by regulating energy metabolism and reducing mitochondrial oxidative stress, thus counteracting the effects of mitochondrial dysfunction in various renal diseases.
  • Ameliorates renal tubular and cardiac myocyte damage
  • Modulates mitochondrial ATP synthesis
  • Protects mitochondrial function
  • Regulates energy metabolism
  • Reduces mitochondrial oxidative stress
  • Inhibits TNFα-induced apoptosis in microglia
  • Improves renal function in ischemia-reperfusion injury and cisplatin-induced nephropathy models
Catalog No. 50-003-47481 Supplier Medchemexpress LLC Supplier No. HY11153650MG
Mitochonic acid 5 (MA-5) is a compound that binds to mitochondria, ameliorating renal tubular and cardiac myocyte damage by modulating mitochondrial ATP synthesis independently of oxidative phosphorylation and the electron transport chain. Derived from indole-3-acetic acid, it protects mitochondrial function by regulating energy metabolism and reducing mitochondrial oxidative stress, thus counteracting the effects of mitochondrial dysfunction in various renal diseases.
  • Ameliorates renal tubular and cardiac myocyte damage
  • Modulates mitochondrial ATP synthesis
  • Protects mitochondrial function
  • Regulates energy metabolism
  • Reduces mitochondrial oxidative stress
  • Inhibits TNFα-induced apoptosis in microglia
  • Improves renal function in ischemia-reperfusion injury and cisplatin-induced nephropathy models