Learn More
Medchemexpress LLC Mitochonic acid 5 | 1354707-41-7 | 99.60% | 329.30 | 25 MG
SDP
Catalog No. 5000346882
Encompass_Preferred
Mitochonic acid 5 (MA-5) is a compound that binds to mitochondria and helps alleviate damage to renal tubules and cardiac myocytes. It modulates mitochondrial ATP synthesis, protecting mitochondrial function by regulating energy metabolism and reducing mitochondrial oxidative stress.
  • Binds mitochondria and ameliorates renal tubular and cardiac myocyte damage.
  • Modulates mitochondrial ATP synthesis.
  • Protects mitochondrial function by regulating energy metabolism and reducing mitochondrial oxidative stress.
  • Dose-dependently inhibits TNFα-induced cell viability decrease in microglia.
  • Improves renal function in ischemia-reperfusion injury and cisplatin-induced nephropathy models.
Catalog No. 50-003-46882 Supplier Medchemexpress LLC Supplier No. 25MGHY111536
Add to Cart
Add to Cart
Mitochonic acid 5 (MA-5) is a compound that binds to mitochondria and helps alleviate damage to renal tubules and cardiac myocytes. It modulates mitochondrial ATP synthesis, protecting mitochondrial function by regulating energy metabolism and reducing mitochondrial oxidative stress.
  • Binds mitochondria and ameliorates renal tubular and cardiac myocyte damage.
  • Modulates mitochondrial ATP synthesis.
  • Protects mitochondrial function by regulating energy metabolism and reducing mitochondrial oxidative stress.
  • Dose-dependently inhibits TNFα-induced cell viability decrease in microglia.
  • Improves renal function in ischemia-reperfusion injury and cisplatin-induced nephropathy models.