Learn More
Cayman Chemical Mitochonic Acid 5, Purity: ≥98%, CAS: 1354707-41-7, 50 mg
SDP
Catalog No. 502910444
Encompass_Preferred
An inhibitor of mitochondrial dysfunction; increases intracellular ATP levels in Hep3B cells at a 3 µM; rescues LPS-induced apoptosis and decreases in the mitochondrial membrane potential in BV-2 microglial cells at 5 µM; protects against BSO-induced decreases in cell viability in fibroblasts isolated from patients with various mitochondrial diseases (EC50s = 2.3-5.1 µM) in an electron transport chain-independent manner; reduces plasma levels of creatinine and decreases acute renal tubular necrosis in a mouse model of bilateral retroperitoneal renal ischemia-reperfusion injury at 50 mg/kg
Catalog No. 50-291-0444 Supplier Cayman Chemical Supplier No. 3570250 MG
An inhibitor of mitochondrial dysfunction; increases intracellular ATP levels in Hep3B cells at a 3 µM; rescues LPS-induced apoptosis and decreases in the mitochondrial membrane potential in BV-2 microglial cells at 5 µM; protects against BSO-induced decreases in cell viability in fibroblasts isolated from patients with various mitochondrial diseases (EC50s = 2.3-5.1 µM) in an electron transport chain-independent manner; reduces plasma levels of creatinine and decreases acute renal tubular necrosis in a mouse model of bilateral retroperitoneal renal ischemia-reperfusion injury at 50 mg/kg