Learn More
Selleck Chemical LLC Mitochonic acid 5 5mg
SDP
Supplier:  Selleck Chemical LLC S08815MG
Encompass_Preferred
Mitochonic acid 5 (MA-5) reduces mitochondrial apoptosis via upregulation of mitophagy. Mitochonic acid 5 regulates mitophagy via Bnip3 through the MAPK-ERK-Yap signaling pathway. Mitochonic acid 5 modulates mitochondrial ATP synthesis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3053