Learn More
Medchemexpress LLC HY-111536 5mg Medchemexpress, Mitochonic acid 5 CAS:1354707-41-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1115365MG
Encompass_Preferred
Medchemexpress, HY-111536 5mg Mitochonic acid 5 CAS:1354707-41-7 Mitochonic acid 5 binds mitochondria and ameliorates renal tubular and cardiac myocyte damage. Mitochonic acid 5 modulates mitochondrial ATP synthesis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8904