Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mitochonic acid 5 (MA-5) | 1354707-41-7 | 99.6% | 329.30 | 1 MG
SDP

Catalog No. 5000347114
Encompass_Preferred

Mitochonic acid 5 (MA-5) binds mitochondria, ameliorating renal tubular and cardiac myocyte damage. It modulates mitochondrial ATP synthesis independently of oxidative phosphorylation and the electron transport chain, protecting mitochondrial function by regulating energy metabolism and reducing oxidative stress. Oral administration has shown increased plasma concentration in a dose-response manner in mice, and it has improved renal function in ischemia-reperfusion injury and cisplatin-induced nephropathy models.

  • Purity: 99.60%
  • Appearance: Solid
  • Color: Light yellow to yellow
  • Molecular weight: 329.30
  • Formula: C18H13F2NO3
  • Solubility (in vitro): ≥ 100 mg/mL in DMSO; insoluble in H2O
  • Storage conditions for powder: -20°C for 3 years, 4°C for 2 years
  • Storage conditions in solvent: -80°C for 2 years, -20°C for 1 year
  • Target: Mitochondrial metabolism

Catalog No. 50-003-47114 Supplier Medchemexpress LLC Supplier No. HY11153610MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Mitochonic acid 5 (MA-5) binds mitochondria, ameliorating renal tubular and cardiac myocyte damage. It modulates mitochondrial ATP synthesis independently of oxidative phosphorylation and the electron transport chain, protecting mitochondrial function by regulating energy metabolism and reducing oxidative stress. Oral administration has shown increased plasma concentration in a dose-response manner in mice, and it has improved renal function in ischemia-reperfusion injury and cisplatin-induced nephropathy models.

  • Purity: 99.60%
  • Appearance: Solid
  • Color: Light yellow to yellow
  • Molecular weight: 329.30
  • Formula: C18H13F2NO3
  • Solubility (in vitro): ≥ 100 mg/mL in DMSO; insoluble in H2O
  • Storage conditions for powder: -20°C for 3 years, 4°C for 2 years
  • Storage conditions in solvent: -80°C for 2 years, -20°C for 1 year
  • Target: Mitochondrial metabolism

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.