Learn More
ABclonal Technology Mitofilin Rabbit mAb
SDP
Catalog No. 5000006141
Encompass_Preferred
Enables RNA binding activity Involved in cristae formation Located in mitochondrial inner membrane Part of MICOS complex
Catalog No. 50-000-06141 Supplier ABclonal Technology Supplier No. A2353020UL
Enables RNA binding activity Involved in cristae formation Located in mitochondrial inner membrane Part of MICOS complex