Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mitogen-activated protein kinase 12 (p38 gamma) | >95.0% | 10 UG
SDP

Catalog No. 5000292612
Encompass_Preferred

Recombinant human p38 gamma (MAPK12) protein expressed in Sf9 insect cells, supplied as a filtered solution for research use. The protein is provided at research-scale quantities, characterized by >95% purity (reducing SDS-PAGE), and shipped on dry ice to maintain cold-chain integrity.

  • Expression host: Sf9 insect cells.
  • Species: human.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Formulated in 20 mM Tris, 500 mM NaCl, 10% glycerol, pH 8.0, 0.2 μm filtered.
  • Tag-free with potential residual N-terminal Gly-Pro.
  • Shipped on dry ice and intended for research use only.

Catalog No. 50-002-92612 Supplier Medchemexpress LLC Supplier No. HYP7653210UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human p38 gamma (MAPK12) protein expressed in Sf9 insect cells, supplied as a filtered solution for research use. The protein is provided at research-scale quantities, characterized by >95% purity (reducing SDS-PAGE), and shipped on dry ice to maintain cold-chain integrity.

  • Expression host: Sf9 insect cells.
  • Species: human.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Formulated in 20 mM Tris, 500 mM NaCl, 10% glycerol, pH 8.0, 0.2 μm filtered.
  • Tag-free with potential residual N-terminal Gly-Pro.
  • Shipped on dry ice and intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.