Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mitogen-activated protein kinase 12 (p38 gamma), human recombinant | >95.0% | 5 UG
SDP

Catalog No. 5000290269
Encompass_Preferred

Recombinant human p38 gamma (MAPK12) protein produced in Sf9 insect cells and supplied as a liquid solution for research use in signaling and kinase assays. Supplied in microgram quantities; store at -80°C for long-term storage and avoid repeated freeze-thaw. Activity is not guaranteed and should be validated by the user prior to functional studies.

  • Recombinant human protein expressed in Sf9 insect cells.
  • Supplied as a ready-to-use liquid solution.
  • Suitable for biochemical assays and antibody validation.
  • Recommended storage at -80°C to preserve stability.
  • Stable at -20°C for short-term storage up to 3 months.
  • Purity reported greater than 95% as determined by reducing SDS-PAGE.
  • Activity not guaranteed; validate prior to use in functional assays.

Catalog No. 50-002-90269 Supplier Medchemexpress LLC Supplier No. HYP765325UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human p38 gamma (MAPK12) protein produced in Sf9 insect cells and supplied as a liquid solution for research use in signaling and kinase assays. Supplied in microgram quantities; store at -80°C for long-term storage and avoid repeated freeze-thaw. Activity is not guaranteed and should be validated by the user prior to functional studies.

  • Recombinant human protein expressed in Sf9 insect cells.
  • Supplied as a ready-to-use liquid solution.
  • Suitable for biochemical assays and antibody validation.
  • Recommended storage at -80°C to preserve stability.
  • Stable at -20°C for short-term storage up to 3 months.
  • Purity reported greater than 95% as determined by reducing SDS-PAGE.
  • Activity not guaranteed; validate prior to use in functional assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.