Learn More
Selleck Chemical LLC Mitoguazone E7797-5MG
SDP
Catalog No. 5000042961
Encompass_Preferred
Mitoguazone is a polyamine analog that disrupts polyamine biosynthesis by inhibiting S-adenosylmethionine decarboxylase (SAMDC) It induces apoptosis and triggers p53-independent programmed cell death in the MCF7 cell line It also inhibits the integration of HIV DNA into cellular DNA in both monocytes and macrophages
Catalog No. 50-000-42961 Supplier Selleck Chemical LLC Supplier No. E77975MG
Mitoguazone is a polyamine analog that disrupts polyamine biosynthesis by inhibiting S-adenosylmethionine decarboxylase (SAMDC) It induces apoptosis and triggers p53-independent programmed cell death in the MCF7 cell line It also inhibits the integration of HIV DNA into cellular DNA in both monocytes and macrophages