Learn More
Medchemexpress LLC Mitoguazone 10mg | 459-86-9 | 10MG
SDP
Catalog No. 5000242148
Encompass_Preferred
Mitoguazone (Methylglyoxal-bis(guanylhydrazone)) is a synthetic polycarbonyl derivative with potent antineoplastic activity Mitoguazone is a brain-penetrant and competitive S-adenosyl-methionine decarboxylase (SAMDC) inhibitor that disrupts polyamine biosynthesis Mitoguazone induces cell apoptosis Mitoguazone inhibits HIV DNA integration into the cellular DNA in both monocytes and macrophages Mitoguazone has the potential for acute leukemia Hodgkin s and non-Hodgkin s lymphoma treatment[1 [2 [3 [4
Catalog No. 50-002-42148 Supplier Medchemexpress LLC Supplier No. HY10663410MG
Mitoguazone (Methylglyoxal-bis(guanylhydrazone)) is a synthetic polycarbonyl derivative with potent antineoplastic activity Mitoguazone is a brain-penetrant and competitive S-adenosyl-methionine decarboxylase (SAMDC) inhibitor that disrupts polyamine biosynthesis Mitoguazone induces cell apoptosis Mitoguazone inhibits HIV DNA integration into the cellular DNA in both monocytes and macrophages Mitoguazone has the potential for acute leukemia Hodgkin s and non-Hodgkin s lymphoma treatment[1 [2 [3 [4