Learn More
Medchemexpress LLC Mitoguazone 5mg | 459-86-9 | 5MG
SDP
Catalog No. 5000240196
Encompass_Preferred
Mitoguazone (Methylglyoxal-bis(guanylhydrazone)) is a synthetic polycarbonyl derivative with potent antineoplastic activity Mitoguazone is a brain-penetrant and competitive S-adenosyl-methionine decarboxylase (SAMDC) inhibitor that disrupts polyamine biosynthesis Mitoguazone induces cell apoptosis Mitoguazone inhibits HIV DNA integration into the cellular DNA in both monocytes and macrophages Mitoguazone has the potential for acute leukemia Hodgkin s and non-Hodgkin s lymphoma treatment[1 [2 [3 [4
Catalog No. 50-002-40196 Supplier Medchemexpress LLC Supplier No. HY1066345MG
Mitoguazone (Methylglyoxal-bis(guanylhydrazone)) is a synthetic polycarbonyl derivative with potent antineoplastic activity Mitoguazone is a brain-penetrant and competitive S-adenosyl-methionine decarboxylase (SAMDC) inhibitor that disrupts polyamine biosynthesis Mitoguazone induces cell apoptosis Mitoguazone inhibits HIV DNA integration into the cellular DNA in both monocytes and macrophages Mitoguazone has the potential for acute leukemia Hodgkin s and non-Hodgkin s lymphoma treatment[1 [2 [3 [4