Learn More
Cayman Chemical MitoLite™ Blue FX490, 500 test
SDP
Catalog No. 502934930
Encompass_Preferred
A mitochondrial dye for live and fixed cells; ex/em = 350/490 nm; has been used to assess mitochondrial localization in NSCLC and SCLC cells
Catalog No. 50-293-4930 Supplier Cayman Chemical Supplier No. 25159500 TEST
A mitochondrial dye for live and fixed cells; ex/em = 350/490 nm; has been used to assess mitochondrial localization in NSCLC and SCLC cells