Learn More
Cayman Chemical MitoLite™ Deep Red FX660, 500 test
SDP
Catalog No. 502934931
Encompass_Preferred
A mitochondrial dye for live and fixed cells; ex/em = 640/659 nm
Catalog No. 50-293-4931 Supplier Cayman Chemical Supplier No. 25160500 TEST
A mitochondrial dye for live and fixed cells; ex/em = 640/659 nm