Learn More
Cayman Chemical MItoLIteGreen EX488 500test
SDP
Catalog No. 502935018
Encompass_Preferred
A mitochondrial dye for live cells; ex/em = 498/520 nm
Catalog No. 50-293-5018 Supplier Cayman Chemical Supplier No. 25161500 TEST
A mitochondrial dye for live cells; ex/em = 498/520 nm