Learn More
Cayman Chemical MItoLIteNIR FX690 500test
SDP
Catalog No. 502933840
Encompass_Preferred
A mitochondrial dye for live or fixed cells; ex/em = 660/692 nm; has been used to determine subcellular localization of photosensitizers modified by polyethylene glycol in HepG2 cells
Catalog No. 50-293-3840 Supplier Cayman Chemical Supplier No. 25163500 TEST
A mitochondrial dye for live or fixed cells; ex/em = 660/692 nm; has been used to determine subcellular localization of photosensitizers modified by polyethylene glycol in HepG2 cells