Learn More
Cayman Chemical MitoLite™ Orange EX405, 500 test
SDP
Catalog No. 502936202
Encompass_Preferred
A mitochondrial dye for live cells; ex/em = 399/550 nm
Catalog No. 50-293-6202 Supplier Cayman Chemical Supplier No. 25164500 TEST
A mitochondrial dye for live cells; ex/em = 399/550 nm