Learn More
Cayman Chemical MitoLite™ Orange FX570, 500 test
SDP
Catalog No. 502936214
Encompass_Preferred
A mitochondrial dye for live and fixed cells; ex/em = 545/575 nm; has been used to label mitochondria in Hep2 cells treated with cisplatin and NAC
Catalog No. 50-293-6214 Supplier Cayman Chemical Supplier No. 25165500 TEST
A mitochondrial dye for live and fixed cells; ex/em = 545/575 nm; has been used to label mitochondria in Hep2 cells treated with cisplatin and NAC