Learn More
Cayman Chemical MitoLite™ Red FX600, 500 test
SDP
Catalog No. 502934933
Encompass_Preferred
A mitochondrial dye for live and fixed cells; ex/em = 575/600 nm
Catalog No. 50-293-4933 Supplier Cayman Chemical Supplier No. 25166500 TEST
A mitochondrial dye for live and fixed cells; ex/em = 575/600 nm