Learn More
Cayman Chemical MItomycIn C 10mg
SDP
Catalog No. 502952100
Encompass_Preferred
An antitumor antibiotic that crosslinks double stranded DNA and is widely used as a tool to selectively inhibit DNA synthesis and mutagenensis, to stimulate genetic recombination, chromosome breakage and sister chromatid exchange, and to induce DNA repair
Catalog No. 50-295-2100 Supplier Cayman Chemical Supplier No. 1143510 MG
An antitumor antibiotic that crosslinks double stranded DNA and is widely used as a tool to selectively inhibit DNA synthesis and mutagenensis, to stimulate genetic recombination, chromosome breakage and sister chromatid exchange, and to induce DNA repair