Learn More
Apexbio Technology LLC Mitomycin C 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC A44525.1
Encompass_Preferred
Mitomycin C (CAS 50-07-7) is an antibiotic compound derived from Streptomyces caespitosus or Streptomyces lavendulae exhibiting anti-cancer properties by interfering with DNA synthesis Its mode of action involves direct formation of covalent adducts with DNA impeding DNA replication In PC3 cells mitomycin C presents an EC50 of approximately 0 14 M Moreover it can potentiate apoptosis induced by TRAIL (TNF-related apoptosis-inducing ligand) through mechanisms independent of p53 modulating apoptosis-related protein expression and caspase activation Due to its cytotoxic mechanism mitomycin C continues to serve as a useful compound in cancer research particularly in apoptosis signaling studies
Catalog No. 50-000-61769