Learn More
Enzo Life Sciences Mitomycin C (2mg). CAS: 50-07-7
SDP
Catalog No. 502010961
Encompass_Preferred
Antitumor antibiotic. DNA crosslinking and damaging agent. Inhibitor of DNA synthesis, nuclear division and cancer cells. DNA intercalating agent. Induces apoptosis in a variety of cells via a p53-dependent pathway. Purity: ≥95% (HPLC). Solubility: Soluble in water (1mg/ml, warm), 100% ethanol (1mg/ml, warm), DMSO (15mg/ml) or methanol. Long Term Storage: -20°C.
This item is not returnable. View return policy
Catalog No. 50-201-0961 Supplier Enzo Life Sciences Supplier No. BMLGR3110002
Antitumor antibiotic. DNA crosslinking and damaging agent. Inhibitor of DNA synthesis, nuclear division and cancer cells. DNA intercalating agent. Induces apoptosis in a variety of cells via a p53-dependent pathway. Purity: ≥95% (HPLC). Solubility: Soluble in water (1mg/ml, warm), 100% ethanol (1mg/ml, warm), DMSO (15mg/ml) or methanol. Long Term Storage: -20°C.