Learn More
Selleck Chemical LLC Mitomycin C
SDP
Catalog No. 5000002975
Encompass_Preferred
Mitomycin C (Ametycine) is an antineoplastic antibiotic by inhibiting DNA synthesis used to treat different cancers Mitomycin C induces apoptosis in a caspases-dependent and Fas/CD95-independent manner This product is a hazardous chemical (acute toxicity/flammable/skin corrosive) Please use it while wearing a protective face mask gloves and clothing
Catalog No. 50-000-02975 Supplier Selleck Chemical LLC Supplier No. S8146200MG
Mitomycin C (Ametycine) is an antineoplastic antibiotic by inhibiting DNA synthesis used to treat different cancers Mitomycin C induces apoptosis in a caspases-dependent and Fas/CD95-independent manner This product is a hazardous chemical (acute toxicity/flammable/skin corrosive) Please use it while wearing a protective face mask gloves and clothing