Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Mitomycin C, contains 2 mg Mitomycin C and 48 mg NaCl

Catalog No. AC226940020
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC226940020 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AC226940020 Supplier Thermo Scientific Chemicals Supplier No. 226940020
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mitomycin C, CAS # 50-07-7, is a methylazirinopyrroloindoledione complex isolated from the bacterium Streptomyces caespitosus and other Streptomyces bacterial species. It is a versatile and potent compound with antibiotic activity and may have potential therapeutic value in the treatment of cancer. | CAS: 50-07-7 | C15H18N4O5 | 334.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

  • Mitomycin C is a water soluble, blue-violet crystalline powder, unstable in light and acidic conditions, but may be stably stored at 2-8°c
  • Bio-reduced mitomycin C generates oxygen radicals, alkylates DNA, and produces inter-strand DNA cross-links, thereby inhibiting DNA synthesis

Applications

  • Mitomycin C is preferentially toxic to hypoxic cells that are resistant to cancer therapy, and is therefore used to study its potential therapeutic effect on advanced stages of cancer
  • At high concentrations, mitomycin C inhibits RNA and protein synthesis and demonstrates strong antibiotic properties towards both Gram-positive and Gram-negative bacteria
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Mitomycin C
CAS 50-07-7, 7647-14-5
Melting Point >360.0°C
Color Blue-Violet
Packaging Ampoule
Molecular Formula C15H18N4O5
Quantity 2 mg
MDL Number MFCD00078109
Merck Index 15, 6300
Synonym mitomycin c, mitomycin, ametycine, mutamycin, ametycin, mitocin-c, mitomycinum, mytomycin, mytozytrex, mitomycin-c
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol, acetone, butyl acetate and, cyclohexanone;, slightly soluble in benzene, ccl4, diethyl ether;, practically insoluble in petroleum ether.
InChI Key NWIBSHFKIJFRCO-WUDYKRTCSA-N
SMILES CC1=C(C(=O)C2=C(C1=O)N3CC4C(C3(C2COC(=O)N)OC)N4)N
Molecular Weight (g/mol) 334.32
PubChem CID 5746
ChEBI CHEBI:27504
Formula Weight 334.32
Percent Purity ≥99% with regard to the non-formulated material
Physical Form Powder
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.