Learn More
Crescent Chemical Co Inc MITOMYCIN C LYPHIL. RESEARCH
SDP
Catalog No. 50247877
Encompass_Preferred
Mitomycin C lyophil. research grade Potent anti-tumor antibiotic isolated from Streptomyces caespitosus. Belongs to the group of aziridine-containing natural products. Causes intra- and interstrand crosslinks in DNA, which prevent dissociation of the strands,
Catalog No. 50-247-877 Supplier Crescent Chemical Co Inc Supplier No. SE29805.01
Mitomycin C lyophil. research grade Potent anti-tumor antibiotic isolated from Streptomyces caespitosus. Belongs to the group of aziridine-containing natural products. Causes intra- and interstrand crosslinks in DNA, which prevent dissociation of the strands,