Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mitomycin C, MP Biomedicals™

Catalog No. MP210049820 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
2 mg
10 mg
20 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP210049820 20 mg
MP210049802 2 mg
MP210049810 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP210049820 Supplier MP Biomedicals, Inc Supplier No. 0210049820
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The most widely used Mitomycin antibiotic. Mitomycin C, MP Biomedicals is the least toxic of the mitomycin antibiotics, with the most potent antitumor activity. It is an inhibitor of DNA synthesis, nuclear division, and cancer cells.

  • Antibacterial to gram positive, gram negative and acid-fast bacilli
  • Decomposes DNA in cancer cells by forming a complex with DNA
  • Inhibits division of cancer cells by interfering with the biosynthesis of DNA
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-07-7
Molecular Formula C15H18N4O5
Molecular Weight (g/mol) 334.332
InChI Key NWIBSHFKIJFRCO-WUDYKRTCSA-N
Synonym Ametycin, 7-Amino-9-a-methoxymitosane, Mit-C, Mitocin-C, Mitomycinum, Mutamycin, Azirizino[2', 3': 3,4] pyrrolo [1,2-a] indole 4,7-dione-6-amino-1,1a, 2, 8, 8a, 8b-hexahydro- 8- (hydroxymethyl) 8a-methoxy-5-methyl carbamate
PubChem CID 5746
ChEBI CHEBI:27504
SMILES CC1=C(C(=O)C2=C(C1=O)N3CC4C(C3(C2COC(=O)N)OC)N4)N

Specifications

Type Mitomycin C
Physical Form Crystalline Powder
pH 6 to 9 (Stability)
Extinction Coefficient EmM(367 nm) = 21.84 (water); EmM(357 nm) = 22.78 (methanol); E1% (216 nm) = 742 (methanol); E1% (360 nm) = 742 (methanol); E1% (560 nm) = 0.06 (methanol) (Lit.)
Potency 850μg/mg minimum
Color Blue
Quantity 20 mg
Solubility Information Soluble in dilute aqueous acids or dilute aqueous bases.
Formula Weight 334.33
Grade Antibiotic
Recommended Storage Store at +4°C.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.