Learn More
Mitomycin C, USP Grade, G-Biosciences

Description
Mitomycin C has been used to produce mitotically inactivated feeder cells and has been shown to increase recombination during fruit fly meiosis. It is capable of killing mutant strains of Arabidopsis that are defective for recombination during meiosis.
Specifications
Specifications
| Form | Crystalline Powder |
| Product Line | Gibco |
| Purity or Quality Grade | Laboratory |
| Quantity | 500 mg |
| Molecular Weight (g/mol) | 334.33 |
| Boiling Point | 581.8°C |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.