Learn More
AdipoGen Mitomycin C (10 mg)
SDP
Supplier:  AdipoGen CDXM0161M010
Encompass_Preferred
Chemical. CAS: 50-07-7. Formula: C15H18N4O5. MW: 334.33. Mitomycin C (MMC) is an antitumor antibiotic. This product is an alkylating agent that specifically targets the guanine nucleoside sequence 5‘-CpG-3‘. It inhibits DNA synthesis by covalently reacting with DNA, forming crosslinks between complementary strands of DNA. This interaction prevents separation of complementary DNA strands, inhibiting DNA replication. Mitomycin C causes the cross-linking of double-stranded DNA, which results in mutagenesis, inhibition of DNA synthesis, initiation of DNA repair events, and activation of apoptosis. In tissue culture, this compound decreases cell viability and suppresses mitosis, and causes disorganization of nucleus and the production of giant cells. Mitomycin C has strong antitumor activity, especially against Ehrlich ascites tumor cells, and strong bactericidal action against gram-positive and gram-negative bacteria.
Catalog No. 50-205-4535