Learn More
Abcam MitoPerOx, fluorescent mitochondria-targeted lipid peroxidation probe
Supplier:  Abcam AB1468201MG
Encompass_Preferred
Ratiometric, fluorescent, mitochondria-targeted lipid peroxidation probe
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2133453