Learn More
Selleck Chemical LLC MitoQ S8978-100MG
SDP
Catalog No. 5000024717
Encompass_Preferred
MitoQ (Mitoquinone) mesylate is a TPP-based mitochondria-targeted antioxidant that blocks H2O2-induced intracellular ROS responses and protects against oxidative damage This product is a waxy solid
Catalog No. 50-000-24717 Supplier Selleck Chemical LLC Supplier No. S8978100MG
MitoQ (Mitoquinone) mesylate is a TPP-based mitochondria-targeted antioxidant that blocks H2O2-induced intracellular ROS responses and protects against oxidative damage This product is a waxy solid