Learn More
Medchemexpress LLC HY-100116A 10mg Medchemexpress, Mitoquinone (mesylate) CAS:845959-50-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY100116A10MG
Encompass_Preferred
Medchemexpress, HY-100116A 10mg Mitoquinone (mesylate) CAS:845959-50-4 Mitoquinone mesylate is a TPP-based, mitochondrially targeted antioxidant in order to protect against oxidative damage. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8609
 Restricted Item