Learn More
Medchemexpress LLC Mitoquinone mesylate | 845959-50-4 | MFCD30747833 | >95.0% | 678.81 g/mol | C38H47O7PS | 25 MG
SDP
Catalog No. 5000215718
Encompass_Preferred
Mitoquinone mesylate is a mitochondria-targeted antioxidant that combines a ubiquinone-derived moiety with a lipophilic triphenylphosphonium (TPP) carrier to concentrate within mitochondria and protect against oxidative damage. It is commonly used in cellular and biochemical studies to decrease mitochondrial reactive oxygen species (ROS) and investigate redox-dependent pathways.
  • Mitochondria-targeted antioxidant that reduces mitochondrial ROS.
  • Accumulation in mitochondria via a lipophilic triphenylphosphonium (TPP) moiety.
  • Suitable for cellular and biochemical studies of oxidative stress and redox biology.
  • Provided as a stable, characterized small-molecule reagent for research use.
  • Documented molecular formula C38H47O7PS and molecular weight 678.81 g/mol.
Catalog No. 50-002-15718 Supplier Medchemexpress LLC Supplier No. HY100116A25MG
Mitoquinone mesylate is a mitochondria-targeted antioxidant that combines a ubiquinone-derived moiety with a lipophilic triphenylphosphonium (TPP) carrier to concentrate within mitochondria and protect against oxidative damage. It is commonly used in cellular and biochemical studies to decrease mitochondrial reactive oxygen species (ROS) and investigate redox-dependent pathways.
  • Mitochondria-targeted antioxidant that reduces mitochondrial ROS.
  • Accumulation in mitochondria via a lipophilic triphenylphosphonium (TPP) moiety.
  • Suitable for cellular and biochemical studies of oxidative stress and redox biology.
  • Provided as a stable, characterized small-molecule reagent for research use.
  • Documented molecular formula C38H47O7PS and molecular weight 678.81 g/mol.