Learn More
Cayman Chemical Mitoquinone (mesylate), Purity: ≥95%, CAS: 845959-50-4, 10 mg
SDP
Catalog No. 502990154
Encompass_Preferred
An oxidized form of mitoquinol; reduced to mitoquinol in isolated bovine heart mitochondrial membranes using succinate for respiration at 50 µM; increases the rate of hydrogen peroxide formation in isolated BAECs at 0.01-10 µM; induces cell death in isolated mouse pancreatic acinar cells at 1 µM; in combination with mitoquinol, reduces MDA levels in isolated rat liver mitochondria and decreases caspase activity in Jurkat cells
Catalog No. 50-299-0154 Supplier Cayman Chemical Supplier No. 2931710 MG
An oxidized form of mitoquinol; reduced to mitoquinol in isolated bovine heart mitochondrial membranes using succinate for respiration at 50 µM; increases the rate of hydrogen peroxide formation in isolated BAECs at 0.01-10 µM; induces cell death in isolated mouse pancreatic acinar cells at 1 µM; in combination with mitoquinol, reduces MDA levels in isolated rat liver mitochondria and decreases caspase activity in Jurkat cells