Learn More
Medchemexpress LLC MitoTam iodide, hydriodide | 1634624-74-0 | 95.7% and 98.8% | 999.82 g·mol⁻¹ | C52H60I2NOP | 10MG
SDP
Catalog No. 5000235870
Encompass_Preferred
MitoTam iodide, hydriodide is a tamoxifen-derived research compound that inhibits the mitochondrial electron transport chain (ETC). It suppresses complex I-dependent respiration, disrupts respiratory supercomplexes, and reduces mitochondrial membrane potential, and is used in preclinical studies of mitochondrial function and anticancer mechanisms. The compound is supplied as the hydriodide (iodide) salt in small milligram quantities for research use.
Catalog No. 50-002-35870 Supplier Medchemexpress LLC Supplier No. HY126222A10MG
MitoTam iodide, hydriodide is a tamoxifen-derived research compound that inhibits the mitochondrial electron transport chain (ETC). It suppresses complex I-dependent respiration, disrupts respiratory supercomplexes, and reduces mitochondrial membrane potential, and is used in preclinical studies of mitochondrial function and anticancer mechanisms. The compound is supplied as the hydriodide (iodide) salt in small milligram quantities for research use.