Learn More
Apexbio Technology LLC Mitotane (Lsodren),100mg CAS# 53-19-0
SDP
Supplier:  Apexbio Technology LLC A8475100
Encompass_Preferred
Mitotane, or o,p'-DDD, is a derivative of DDT with antiproliferative properties. A study of the antiproliferative effects of Mitotane demonstrated particular efficacy against adrenocortical carcinoma. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-224-0303