Learn More
Apexbio Technology LLC Mitotane (Lsodren) 53-19-0 10mM (in 1mL DMSO)
SDP
Catalog No. 5000061782
Encompass_Preferred
Mitotane (Lysodren CAS 53-19-0) chemically known as o p -DDD is a structural analog of the pesticide DDT with established antiproliferative activity Its primary biological activity involves selective impairment of adrenocortical steroidogenesis resulting in cytotoxicity to adrenal cortical cells Mitotane is utilized predominantly in research protocols exploring therapeutic interventions for adrenocortical carcinoma a malignancy arising from adrenal cortical tissue
Catalog No. 50-000-61782 Supplier Apexbio Technology LLC Supplier No. A84755.1
Mitotane (Lysodren CAS 53-19-0) chemically known as o p -DDD is a structural analog of the pesticide DDT with established antiproliferative activity Its primary biological activity involves selective impairment of adrenocortical steroidogenesis resulting in cytotoxicity to adrenal cortical cells Mitotane is utilized predominantly in research protocols exploring therapeutic interventions for adrenocortical carcinoma a malignancy arising from adrenal cortical tissue