Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mitotane

Catalog No. AC109850050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC109850050 5 g
AC109850010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC109850050 Supplier Thermo Scientific Chemicals Supplier No. 109850050

CAS: 53-19-0 | C14H10Cl4 | 320.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53-19-0
Molecular Formula C14H10Cl4
Molecular Weight (g/mol) 320.03
InChI Key JWBOIMRXGHLCPP-UHFFFAOYNA-N
IUPAC Name 1-chloro-4-[2,2-dichloro-1-(2-chlorophenyl)ethyl]benzene
SMILES ClC(Cl)C(C1=CC=C(Cl)C=C1)C1=CC=CC=C1Cl

Specifications

Melting Point 77.0°C to 78.0°C
Linear Formula (ClC6H4)2CHCHCl2
Quantity 5 g
Merck Index 15, 6301
Solubility Information Solubility aq. soln.: .. Other solubilities: soluble in dmso, methanol, ethanol, isooctane and, ccl4
Formula Weight 320.04
Chemical Name or Material Mitotane
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Suspected of causing cancer.

Precautionary Statement GHS P Statement Obtain special instructions before use.

Wear protective gloves/protective clothing/eye protection/face protection.

IF exposed or concerned: Get medical advice/attention.

EINECSNumber 200-166-6

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.