Learn More
Cayman Chemical MItoTEMPOhydrate 10mg
SDP
Catalog No. 502980564
Encompass_Preferred
A mitochondria-targeted superoxide dismutase mimetic that possesses superoxide and alkyl radical scavenging properties
Catalog No. 50-298-0564 Supplier Cayman Chemical Supplier No. 1662110 MG
A mitochondria-targeted superoxide dismutase mimetic that possesses superoxide and alkyl radical scavenging properties