Learn More
Abcam MitoTEMPOL, mitochondria-targeted antio x idant agent, 25MG
Catalog No. 502705534
Encompass_Preferred
MW 583.6 Da, Purity >98%. Mitochondria-targeted antioxidant agent. SOD mimetic Tempol derivative. Protects complex I and II/III respiration. Restores MnSOD activity. Shows cardioprotective effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5534 Supplier Abcam Supplier No. AB14464425MG
MW 583.6 Da, Purity >98%. Mitochondria-targeted antioxidant agent. SOD mimetic Tempol derivative. Protects complex I and II/III respiration. Restores MnSOD activity. Shows cardioprotective effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement