Learn More
Cell Signaling Technology MitoTracker Green FM, Live Cell Imaging, 500 μg
SDP
Catalog No. 502050582
Encompass
MitoTracker™ Green FM is a cell-permeable probe designed for labeling mitochondria in live cells. It contains a mildly thiol-reactive chloromethyl moiety that allows the dye to accumulate in active mitochondria by diffusing across the plasma membrane. It is specifically optimized for live-cell imaging; subsequent fixation with aldehydes or alcohols will inhibit staining.
  • Excitation and emission wavelengths of 490 nm and 516 nm, respectively.
  • Supplied as 50 μg lyophilized solid per vial.
  • Reconstitute with 74.4 μl high-quality DMSO to produce a 1 mM stock solution.
  • Recommended staining concentration is 100-400 nM with incubation for 15-30 min at 37°C.
  • Compatible with phenol red-free media to minimize background fluorescence.
  • Suitable for immunofluorescence and flow cytometry applications.
Catalog No. 50-205-0582 Supplier Cell Signaling Technology Supplier No. 9074S
MitoTracker™ Green FM is a cell-permeable probe designed for labeling mitochondria in live cells. It contains a mildly thiol-reactive chloromethyl moiety that allows the dye to accumulate in active mitochondria by diffusing across the plasma membrane. It is specifically optimized for live-cell imaging; subsequent fixation with aldehydes or alcohols will inhibit staining.
  • Excitation and emission wavelengths of 490 nm and 516 nm, respectively.
  • Supplied as 50 μg lyophilized solid per vial.
  • Reconstitute with 74.4 μl high-quality DMSO to produce a 1 mM stock solution.
  • Recommended staining concentration is 100-400 nM with incubation for 15-30 min at 37°C.
  • Compatible with phenol red-free media to minimize background fluorescence.
  • Suitable for immunofluorescence and flow cytometry applications.