Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cell Signaling Technology MitoTracker Red CMXRos, Cellular Dye, 500 μg
SDP

Catalog No. NC1130609
Encompass

MitoTracker™ Red CMXRos is a cell-permeable probe containing a mildly thiol-reactive chloromethyl moiety designed for mitochondrial labeling. It is well-retained after fixation, allowing for subsequent sample processing and immunostaining. The dye passively diffuses across the plasma membrane and accumulates in active mitochondria.

  • Excitation: 579 nm; Emission: 599 nm
  • Molecular weight: 531.52
  • Validated applications include immunofluorescence (immunocytochemistry) and flow cytometry
  • Retained after fixation with ice-cold methanol
  • Species reactivity: all species expected
  • Store lyophilized solid at -20°C, desiccated and protected from light
  • Reconstitute in DMSO to create a 1 mM stock solution
  • Typical staining concentration: 50-200 nM
  • Optimal incubation: 15-45 min at 37°C

Catalog No. NC1130609 Supplier Cell Signaling Technology Supplier No. 9082S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MitoTracker™ Red CMXRos is a cell-permeable probe containing a mildly thiol-reactive chloromethyl moiety designed for mitochondrial labeling. It is well-retained after fixation, allowing for subsequent sample processing and immunostaining. The dye passively diffuses across the plasma membrane and accumulates in active mitochondria.

  • Excitation: 579 nm; Emission: 599 nm
  • Molecular weight: 531.52
  • Validated applications include immunofluorescence (immunocytochemistry) and flow cytometry
  • Retained after fixation with ice-cold methanol
  • Species reactivity: all species expected
  • Store lyophilized solid at -20°C, desiccated and protected from light
  • Reconstitute in DMSO to create a 1 mM stock solution
  • Typical staining concentration: 50-200 nM
  • Optimal incubation: 15-45 min at 37°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.