Learn More
Biotium MitoView™ 405 (Sample size)
Catalog No. 501964774
Encompass_Preferred
MitoView™ 405 is a blue fluorescent mitochondrial dye for live cells. It has excitation/emission at 398/440 nm for detection in the DAPI or Pacific Blue channels. It is a replacement for our original MitoView™ Blue, with improved photostability.
Catalog No. 50-196-4774 Supplier Biotium Supplier No. 70070T
MitoView™ 405 is a blue fluorescent mitochondrial dye for live cells. It has excitation/emission at 398/440 nm for detection in the DAPI or Pacific Blue channels. It is a replacement for our original MitoView™ Blue, with improved photostability.