Learn More
Biotium MitoView™ 650
Catalog No. 501964775
Encompass_Preferred
MitoView™ 650 is a far-red mitochondrial stain for live cells. Its staining is not responsive to mitochondrial membrane potential. With excitation/emission at 644/670 nm, and can be combined with visible red probes for multicolor imaging.
Catalog No. 50-196-4775 Supplier Biotium Supplier No. 70075
MitoView™ 650 is a far-red mitochondrial stain for live cells. Its staining is not responsive to mitochondrial membrane potential. With excitation/emission at 644/670 nm, and can be combined with visible red probes for multicolor imaging.