Learn More
Biotium MitoView™ 720, 1000X in DMSO
Catalog No. 501964773
Encompass_Preferred
MitoView™ 720 is a near-infrared live cell mitochondrial dye with absorbance/emission at 720/758 nm. While optimally detected using Cy7 settings, the dye is bright enough to be imaged in the Cy5 channel.
Catalog No. 50-196-4773 Supplier Biotium Supplier No. 70068
MitoView™ 720 is a near-infrared live cell mitochondrial dye with absorbance/emission at 720/758 nm. While optimally detected using Cy7 settings, the dye is bright enough to be imaged in the Cy5 channel.